Assay Details
| Gene Symbol: TRIB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001111877.4 | XP_001111877.2 | 864 | 120 | ||
| XM_002798180.3 | XP_002798226.2 | 766 | 120 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRIB3| Gene Symbol: TRIB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001111877.4 | XP_001111877.2 | 864 | 120 | ||
| XM_002798180.3 | XP_002798226.2 | 766 | 120 | |||
Set Membership: |
|