Assay Details
| Gene Symbol: SQOR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001112121.4 | XP_001112121.2 | 1226 | 132 | ||
| XM_015142149.2 | XP_014997635.2 | 965 | 132 | |||
| XM_028850878.1 | XP_028706711.1 | 1296 | 132 | |||
| XM_028850879.1 | XP_028706712.1 | 675 | 132 | |||
| GenBank mRNA | JU326303.1 | - | 729 | 132 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SQOR
Genomic Map