Assay Details
| Gene Symbol: SQOR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001112121.4 | XP_001112121.2 | 1424 | 144 | ||
| XM_015142149.2 | XP_014997635.2 | 1163 | 144 | |||
| XM_028850878.1 | XP_028706711.1 | 1494 | 144 | |||
| XM_028850879.1 | XP_028706712.1 | 873 | 144 | |||
More Information
Set Membership: |
|

SQOR
Genomic Map