Assay Details
| Gene Symbol: MS4A7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001261147.1 | NP_001248076.1 | 491 | 121 | ||
| XM_015114052.2 | XP_014969538.1 | 535 | 121 | |||
| GenBank mRNA | JU323433.1 | - | 491 | 121 | ||
| JV490267.1 | - | 369 | 121 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MS4A7
Genomic Map