Assay Details
| Gene Symbol: OVOL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015149546.2 | XP_015005032.1 | 1623 | 90 | ||
| XM_015149548.2 | XP_015005034.1 | 1629 | 90 | |||
| XM_028828160.1 | XP_028683993.1 | 1510 | 90 | |||
| GenBank mRNA | JV477128.1 | - | 44 | 90 | ||
| JV601601.1 | - | 566 | 90 | |||
| JV667300.1 | - | 417 | 90 | |||
More Information
Set Membership: |
|

OVOL2
Genomic Map