Assay Details
| Gene Symbol: DNASE2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001106353.3 | XP_001106353.1 | 618 | 130 | ||
| XM_015144139.2 | XP_014999625.1 | 620 | 130 | |||
More Information
Set Membership: |
|

DNASE2B
Genomic Map