Assay Details
| Gene Symbol: EDA2R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193467.1 | NP_001180396.1 | 267 | 79 | ||
| XM_015127601.2 | XP_014983087.1 | 357 | 79 | |||
| XM_028841807.1 | XP_028697640.1 | 375 | 79 | |||
| XM_028841808.1 | XP_028697641.1 | 375 | 79 | |||
| XM_028841809.1 | XP_028697642.1 | 357 | 79 | |||
More Information
Set Membership: |
|

EDA2R
Genomic Map