Assay Details
| Gene Symbol: IL21R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125835.2 | XP_014981321.1 | 990 | 99 | ||
| GenBank mRNA | JU478336.1 | - | 956 | 99 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

IL21R
Genomic Map