Assay Details
| Gene Symbol: ZFAND3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015136277.2 | XP_014991763.1 | 1028 | 136 | ||
| XM_015136278.2 | XP_014991764.1 | 980 | 136 | |||
| XM_015136279.2 | XP_014991765.1 | 914 | 136 | |||
| GenBank mRNA | JU335811.1 | - | 632 | 136 | ||
| JU470464.1 | - | 616 | 136 | |||
| JV047862.1 | - | 571 | 136 | |||
| JV553853.1 | - | 645 | 136 | |||
| JV658047.1 | - | 643 | 136 | |||
More Information
Set Membership: |
|

ZFAND3
Genomic Map