Assay Details
| Gene Symbol: C7H14orf93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028851406.1 | XP_028707239.1 | 1682 | 140 | ||
| GenBank mRNA | JU326304.1 | - | 812 | 140 | ||
| JU473767.1 | - | 1470 | 140 | |||
More Information
Set Membership: |
|

C7H14orf93
Genomic Map