Assay Details
| Gene Symbol: MCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266501.2 | NP_001253430.1 | 707 | 116 | ||
| GenBank mRNA | JU323004.1 | - | 752 | 116 | ||
| JU470857.1 | - | 817 | 116 | |||
| JU470858.1 | - | 817 | 116 | |||
| JV047075.1 | - | 707 | 116 | |||
More Information
Set Membership: |
|

MCL1
Genomic Map