Assay Details
| Gene Symbol: MCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266501.2 | NP_001253430.1 | 952 | 104 | ||
| GenBank mRNA | JU470857.1 | - | 1062 | 104 | ||
| JU470858.1 | - | 1062 | 104 | |||
| JV047075.1 | - | 952 | 104 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MCL1
Genomic Map