Assay Details
| Gene Symbol: EDAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015113103.2 | XP_014968589.2 | 965 | 71 | ||
| XM_028832289.1 | XP_028688122.1 | 945 | 71 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EDAR| Gene Symbol: EDAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015113103.2 | XP_014968589.2 | 965 | 71 | ||
| XM_028832289.1 | XP_028688122.1 | 945 | 71 | |||
Set Membership: |
|