Assay Details
| Gene Symbol: GLIPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266729.1 | NP_001253658.1 | 347 | 123 | ||
| GenBank mRNA | JU322063.1 | - | 342 | 123 | ||
| JV046276.1 | - | 347 | 123 | |||
More Information
Set Membership: |
|

GLIPR2
Genomic Map