Assay Details
| Gene Symbol: DPEP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001097323.4 | XP_001097323.1 | 693 | 70 | ||
| XM_028841539.1 | XP_028697372.1 | 1534 | 70 | |||
| XM_028841540.1 | XP_028697373.1 | 693 | 70 | |||
| XM_028841541.1 | XP_028697374.1 | 210 | 70 | |||
| GenBank mRNA | JU491464.1 | - | 197 | 70 | ||
More Information
Set Membership: |
|

DPEP2
Genomic Map