Assay Details
| Gene Symbol: CIAO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204755.1 | NP_001191684.1 | 487 | 93 | ||
| GenBank mRNA | DV769062.1 | - | 109 | 93 | ||
| JU335923.1 | - | 444 | 93 | |||
| JU476804.1 | - | 455 | 93 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CIAO3
Genomic Map