Assay Details
| Gene Symbol: CIAO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204755.1 | NP_001191684.1 | 1079 | 80 | ||
| GenBank mRNA | JU335923.1 | - | 1036 | 80 | ||
| JU476804.1 | - | 1047 | 80 | |||
More Information
Set Membership: |
|

CIAO3
Genomic Map