Assay Details
| Gene Symbol: DUSP26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015145054.2 | XP_015000540.1 | 954 | 138 | ||
| XM_028852126.1 | XP_028707959.1 | 960 | 138 | |||
| GenBank mRNA | JV453105.1 | - | 747 | 138 | ||
More Information
Set Membership: |
|

DUSP26
Genomic Map