Assay Details
| Gene Symbol: AUNIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001107799.4 | XP_001107799.3 | 322 | 151 | ||
| XM_028837837.1 | XP_028693670.1 | 442 | 151 | |||
| XM_028837842.1 | XP_028693675.1 | 348 | 151 | |||
| XM_028837851.1 | XP_028693684.1 | 593 | 151 | |||
| XM_028837852.1 | XP_028693685.1 | 783 | 151 | |||
| GenBank mRNA | JU493203.1 | - | 142 | 151 | ||
More Information
Set Membership: |
|

AUNIP
Genomic Map