Assay Details
| Gene Symbol: GGCT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266969.1 | NP_001253898.1 | 418 | 161 | ||
| GenBank mRNA | JU321051.1 | - | 418 | 161 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GGCT| Gene Symbol: GGCT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266969.1 | NP_001253898.1 | 418 | 161 | ||
| GenBank mRNA | JU321051.1 | - | 418 | 161 | ||
Set Membership: |
|