Assay Details
Gene Symbol: POGLUT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015121421.2 | XP_014976907.1 | 1493 | 126 | ||
GenBank mRNA | JU336344.1 | - | 1208 | 126 | ||
JV045557.1 | - | 1332 | 126 | |||
JV661190.1 | - | 1392 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
