Assay Details
| Gene Symbol: RETREG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193956.2 | NP_001180885.2 | 682 | 89 | ||
| GenBank mRNA | JU326611.1 | - | 534 | 89 | ||
| JV527814.1 | - | 557 | 89 | |||
| JV557589.1 | - | 557 | 89 | |||

RETREG2
Genomic Map