Assay Details
Gene Symbol: C19H19orf57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028839473.1 | XP_028695306.1 | 98 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028839473.1 transcripts.
Set Membership: |
![]() ![]() |
