Assay Details
| Gene Symbol: C19H19orf57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001111192.4 | XP_001111192.4 | 391 | 153 | ||
| XM_015123105.2 | XP_014978591.2 | 378 | 153 | |||
| XM_015123106.2 | XP_014978592.2 | 757 | 153 | |||
| XM_028839473.1 | XP_028695306.1 | 399 | 153 | |||
More Information
Set Membership: |
|

C19H19orf57
Genomic Map