Assay Details
| Gene Symbol: PAGR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194289.2 | NP_001181218.2 | 665 | 127 | ||
| GenBank mRNA | JU336376.1 | - | 814 | 127 | ||
| JU477193.1 | - | 814 | 127 | |||
| JV048244.1 | - | 716 | 127 | |||
| JV661329.1 | - | 814 | 127 | |||
More Information
Set Membership: |
|

PAGR1
Genomic Map