Assay Details
| Gene Symbol: ADIPOR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266618.1 | NP_001253547.1 | 681 | 89 | ||
| XM_028828900.1 | XP_028684733.1 | 842 | 89 | |||
| XM_028828901.1 | XP_028684734.1 | 683 | 89 | |||
| XM_028828902.1 | XP_028684735.1 | 483 | 89 | |||
| GenBank mRNA | JU336383.1 | - | 559 | 89 | ||
| JU473640.1 | - | 549 | 89 | |||
| JV048251.1 | - | 681 | 89 | |||
| JV638645.1 | - | 630 | 89 | |||
More Information
Set Membership: |
|

ADIPOR2
Genomic Map