Assay Details
| Gene Symbol: ZDHHC14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266260.1 | NP_001253189.1 | 278 | 109 | ||
| XM_015137052.2 | XP_014992538.1 | 1026 | 109 | |||
| XM_015137053.2 | XP_014992539.1 | 254 | 109 | |||
| XM_028846700.1 | XP_028702533.1 | 254 | 109 | |||
| GenBank mRNA | JU336404.1 | - | 278 | 109 | ||
| JU336405.1 | - | 278 | 109 | |||
| JV557973.1 | - | 305 | 109 | |||
| JV593749.1 | - | 301 | 109 | |||
More Information
Set Membership: |
|

ZDHHC14
Genomic Map