Assay Details
| Gene Symbol: NSUN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015138232.2 | XP_014993718.2 | 1358 | 89 | ||
| XM_015138233.2 | XP_014993719.1 | 1358 | 89 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NSUN7| Gene Symbol: NSUN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015138232.2 | XP_014993718.2 | 1358 | 89 | ||
| XM_015138233.2 | XP_014993719.1 | 1358 | 89 | |||
Set Membership: |
|