Assay Details
| Gene Symbol: ECHDC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001082412.4 | XP_001082412.1 | 402 | 145 | ||
| GenBank mRNA | JV628915.1 | - | 381 | 145 | ||
| JV637543.1 | - | 145 | ||||
More Information
Set Membership: |
|

ECHDC3
Genomic Map