Assay Details
| Gene Symbol: IQCA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015111397.2 | XP_014966883.2 | 1767 | 96 | ||
| XM_015111398.2 | XP_014966884.2 | 1764 | 96 | |||
| XM_015111399.2 | XP_014966885.2 | 1575 | 96 | |||
| XM_028831259.1 | XP_028687092.1 | 1768 | 96 | |||
| GenBank mRNA | JV723560.1 | - | 379 | 96 | ||
More Information
Set Membership: |
|

IQCA1
Genomic Map