Assay Details
| Gene Symbol: ZMYM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015134981.2 | XP_014990467.2 | 283 | 92 | ||
| XM_015134988.2 | XP_014990474.2 | 314 | 92 | |||
| XM_028836147.1 | XP_028691980.1 | 606 | 92 | |||
| XM_028836156.1 | XP_028691989.1 | 150 | 92 | |||
| XM_028836162.1 | XP_028691995.1 | 606 | 92 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028836156.1, XM_028836162.1 transcripts.
Set Membership: |
|

ZMYM1
Genomic Map