Assay Details
Gene Symbol: TLCD3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028835838.1 | XP_028691671.1 | 319 | 134 | ||
GenBank mRNA | JU493664.1 | - | 89 | 134 |

More Information
Set Membership: |
![]() ![]() ![]() |
