Assay Details
| Gene Symbol: BAALC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266643.1 | NP_001253572.1 | 498 | 119 | ||
| GenBank mRNA | JU336438.1 | - | 498 | 119 | ||
| JV558302.1 | - | 523 | 119 | |||
More Information
Set Membership: |
|

BAALC
Genomic Map