Assay Details
| Gene Symbol: RABEP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125856.2 | XP_014981342.1 | 2077 | 61 | ||
| GenBank mRNA | JV048281.1 | - | 1521 | 61 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RABEP2| Gene Symbol: RABEP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125856.2 | XP_014981342.1 | 2077 | 61 | ||
| GenBank mRNA | JV048281.1 | - | 1521 | 61 | ||
Set Membership: |
|