Assay Details
| Gene Symbol: CSRNP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257746.1 | NP_001244675.1 | 392 | 116 | ||
| XM_015110246.1 | XP_014965732.1 | 531 | 116 | |||
| XM_015110248.2 | XP_014965734.1 | 346 | 116 | |||
| GenBank mRNA | JU322251.1 | - | 392 | 116 | ||
More Information
Set Membership: |
|

CSRNP3
Genomic Map