Assay Details
| Gene Symbol: TEDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193760.2 | NP_001180689.2 | 231 | 142 | ||
| XM_015125328.2 | XP_014980814.2 | 625 | 142 | |||
| GenBank mRNA | JU493918.1 | - | 227 | 142 | ||
More Information
Set Membership: |
|

TEDC2
Genomic Map