Assay Details
| Gene Symbol: TEDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193760.2 | NP_001180689.2 | 1002 | 87 | ||
| XM_015125328.2 | XP_014980814.2 | 1396 | 87 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TEDC2| Gene Symbol: TEDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193760.2 | NP_001180689.2 | 1002 | 87 | ||
| XM_015125328.2 | XP_014980814.2 | 1396 | 87 | |||
Set Membership: |
|