Assay Details
| Gene Symbol: EFHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257942.1 | NP_001244871.1 | 761 | 107 | ||
| GenBank mRNA | JU336498.1 | - | 761 | 107 | ||
| JV521364.1 | - | 395 | 107 | |||
More Information
Set Membership: |
|

EFHD1
Genomic Map