Assay Details
Gene Symbol: HM13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257420.1 | NP_001244349.1 | 941 | 74 | ||
XM_015149241.2 | XP_015004727.1 | 1039 | 74 | |||
XM_028827358.1 | XP_028683191.1 | 1168 | 74 | |||
XM_028827359.1 | XP_028683192.1 | 1168 | 74 | |||
GenBank mRNA | JU322258.1 | - | 941 | 74 | ||
JU336534.1 | - | 941 | 74 | |||
JU477350.1 | - | 937 | 74 | |||
JV048335.1 | - | 961 | 74 | |||
JV528758.1 | - | 957 | 74 | |||
JV662517.1 | - | 942 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
