Assay Details
Gene Symbol: VOPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015133438.2 | XP_014988924.1 | 305 | 105 | ||
GenBank mRNA | JU321438.1 | - | 132 | 105 | ||
JV043239.1 | - | 163 | 105 | |||
JV601826.1 | - | 201 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() |
