Assay Details
| Gene Symbol: THAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193501.2 | NP_001180430.1 | 779 | 165 | ||
| GenBank mRNA | JU337410.1 | - | 474 | 165 | ||
| JU494500.1 | - | 610 | 165 | |||
| JV724632.1 | - | 594 | 165 | |||
More Information
Set Membership: |
|

THAP2
Genomic Map