Assay Details
| Gene Symbol: RASSF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001087653.4 | XP_001087653.1 | 1089 | 106 | ||
| XM_015118030.2 | XP_014973516.1 | 929 | 106 | |||
| GenBank mRNA | JU320335.1 | - | 654 | 106 | ||
| JU327304.1 | - | 531 | 106 | |||
| JU472887.1 | - | 920 | 106 | |||
| JV662897.1 | - | 676 | 106 | |||
More Information
Set Membership: |
|

RASSF5
Genomic Map