Assay Details
| Gene Symbol: GRWD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001113650.4 | XP_001113650.2 | 889 | 96 | ||
| GenBank mRNA | JU324091.1 | - | 306 | 96 | ||
| JU472394.1 | - | 306 | 96 | |||
| JV048398.1 | - | 306 | 96 | |||
| JV662986.1 | - | 337 | 96 | |||
More Information
Set Membership: |
|

GRWD1
Genomic Map