Assay Details
| Gene Symbol: PXDNL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015145355.2 | XP_015000841.2 | 5948 | 123 | ||
| XM_015145356.2 | XP_015000842.2 | 4617 | 123 | |||
| XM_015145357.2 | XP_015000843.2 | 5681 | 123 | |||
| XM_015145358.2 | XP_015000844.2 | 3539 | 123 | |||
| XM_015145359.2 | XP_015000845.2 | 3898 | 123 | |||
More Information
Set Membership: |
|

PXDNL
Genomic Map