Assay Details
Gene Symbol: LOC713879 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001105834.3 | XP_001105834.1 | 983 | 68 | ||
XM_015123623.2 | XP_014979109.1 | 546 | 68 | |||
XM_015123624.2 | XP_014979110.1 | 159 | 68 | |||
XM_015123625.2 | XP_014979111.1 | 208 | 68 | |||
XM_028839052.1 | XP_028694885.1 | 236 | 68 | |||
XM_028839053.1 | XP_028694886.1 | 983 | 68 | |||
GenBank mRNA | JV052696.1 | - | 195 | 68 | ||
JV643217.1 | - | 195 | 68 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001105834.3, XM_015123623.2, XM_015123624.2, XM_015123625.2, XM_028839052.1, XM_028839053.1 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
