Assay Details
| Gene Symbol: LOC713879 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001105834.3 | XP_001105834.1 | 1118 | 134 | ||
| XM_015123623.2 | XP_014979109.1 | 681 | 134 | |||
| XM_015123624.2 | XP_014979110.1 | 294 | 134 | |||
| XM_015123625.2 | XP_014979111.1 | 343 | 134 | |||
| XM_028839052.1 | XP_028694885.1 | 371 | 134 | |||
| XM_028839053.1 | XP_028694886.1 | 1118 | 134 | |||
| GenBank mRNA | JV052696.1 | - | 330 | 134 | ||
| JV643217.1 | - | 330 | 134 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028839053.1 transcripts.
Set Membership: |
|

LOC713879
Genomic Map