Assay Details
| Gene Symbol: C4H6orf136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105163.1 | NP_001098633.1 | 340 | 77 | ||
| GenBank mRNA | JU333407.1 | - | 883 | 77 | ||
| JV637207.1 | - | 878 | 77 | |||
More Information
Set Membership: |
|

C4H6orf136
Genomic Map