Assay Details
| Gene Symbol: NXPE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194109.2 | NP_001181038.1 | 1629 | 72 | ||
| XM_015128755.2 | XP_014984241.1 | 1858 | 72 | |||
| XM_015128760.2 | XP_014984246.1 | 1824 | 72 | |||
| XM_015128764.2 | XP_014984250.1 | 1456 | 72 | |||
| XM_015128769.2 | XP_014984255.1 | 1581 | 72 | |||
| XM_028843194.1 | XP_028699027.1 | 1635 | 72 | |||
| GenBank mRNA | JU337073.1 | - | 1447 | 72 | ||
| JU337074.1 | - | 1608 | 72 | |||
| JU477804.1 | - | 1301 | 72 | |||
| JV048672.1 | - | 1078 | 72 | |||
More Information
Set Membership: |
|

NXPE3
Genomic Map