Assay Details
| Gene Symbol: DNAJC19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266267.1 | NP_001253196.1 | 302 | 150 | ||
| GenBank mRNA | JU320144.1 | - | 295 | 150 | ||
| JU472559.1 | - | 302 | 150 | |||
More Information
Set Membership: |
|

DNAJC19
Genomic Map