Assay Details
| Gene Symbol: FAM122B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015128231.2 | XP_014983717.1 | 1123 | 73 | ||
| XM_015128232.2 | XP_014983718.1 | 246 | 73 | |||
| XM_015128233.2 | XP_014983719.1 | 1123 | 73 | |||
| XM_015128234.2 | XP_014983720.1 | 246 | 73 | |||
| XM_015128235.2 | XP_014983721.1 | 1123 | 73 | |||
| XM_015128236.2 | XP_014983722.1 | 1123 | 73 | |||
| XM_015128237.2 | XP_014983723.1 | 1030 | 73 | |||
| XM_015128238.2 | XP_014983724.1 | 1123 | 73 | |||
| XM_015128239.2 | XP_014983725.1 | 1123 | 73 | |||
| XM_015128240.2 | XP_014983726.1 | 1123 | 73 | |||
| XM_015128242.2 | XP_014983728.1 | 246 | 73 | |||
| XM_015128243.2 | XP_014983729.1 | 246 | 73 | |||
| XM_015128244.2 | XP_014983730.1 | 246 | 73 | |||
| XM_028842429.1 | XP_028698262.1 | 246 | 73 | |||
| XM_028842430.1 | XP_028698263.1 | 246 | 73 | |||
| GenBank mRNA | JU333150.1 | - | 211 | 73 | ||
| JU333151.1 | - | 211 | 73 | |||
| JU333152.1 | - | 211 | 73 | |||
| JU474478.1 | - | 217 | 73 | |||
| JU474479.1 | - | 217 | 73 | |||
| JU474480.1 | - | 217 | 73 | |||
| JV046294.1 | - | 249 | 73 | |||
| JV046295.1 | - | 249 | 73 | |||
| JV046296.1 | - | 249 | 73 | |||
| JV635984.1 | - | 243 | 73 | |||
More Information
Set Membership: |
|

FAM122B
Genomic Map